KEY428075853 Key Organics / BIONET

4-(propan-2-yloxy)cyclohexan-1-one

  • MDL number MFCD16658578
  • Cas Number : 69697-46-7
  • Molecular Weight : 156.225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4114_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart