KEY428075856 Key Organics / BIONET

5-methyl-1,2-benzoxazol-3-amine

  • MDL number MFCD09907911
  • Cas Number : 89976-56-7
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1,2-benzisoxazol-3-amine
Inchi Code 1S/C8H8N2O/c1-5-2-3-7-6(4-5)8(9)10-11-7/h2-4H,1H3,(H2,9,10)
Inchi Key GNWQLQVKOQHMPF-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436427

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4119_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart