KEY428075857 Key Organics / BIONET

2-fluorobenzene-1,4-dicarbonitrile

  • MDL number MFCD01657004
  • Cas Number : 1897-53-6
  • Molecular Weight : 146.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoroterephthalonitrile
Inchi Code 1S/C8H3FN2/c9-8-3-6(4-10)1-2-7(8)5-11/h1-3H
Inchi Key DULBJBXBNLVFBI-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436428

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4120_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart