KEY428075863 Key Organics / BIONET

2-(4-chloro-3-methoxyphenyl)acetonitrile

  • MDL number MFCD09907926
  • Cas Number : 13726-21-1
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436434

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4126_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart