KEY428075871 Key Organics / BIONET

1-(4-bromo-2-methoxyphenyl)-4-methyl-1H-imidazole

  • MDL number MFCD11040217
  • Cas Number : 870838-56-5
  • Molecular Weight : 267.126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436442

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4137_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart