KEY428075874 Key Organics / BIONET

4-amino-3-ethoxybenzamide

  • MDL number MFCD18338358
  • Cas Number : 917909-47-8
  • Molecular Weight : 180.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436445

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4182_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart