KEY428075876 Key Organics / BIONET

2-bromo-4-fluoro-5-methoxybenzoic acid

  • MDL number MFCD13193242
  • Cas Number : 1007455-21-1
  • Molecular Weight : 249.035

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436447

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4186_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart