KEY428075878 Key Organics / BIONET

2-bromo-5-chloropyridine-4-carboxylic acid

  • MDL number MFCD13181649
  • Cas Number : 530156-90-2
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4188_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart