KEY428075880 Key Organics / BIONET

2-chloro-5,6,7,8-tetrahydro-1,6-naphthyridin-5-one

  • MDL number MFCD16658783
  • Cas Number : 1226898-93-6
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-7,8-dihydro[1,6]naphthyridin-5(6H)-one
Inchi Code 1S/C8H7ClN2O/c9-7-2-1-5-6(11-7)3-4-10-8(5)12/h1-2H,3-4H2,(H,10,12)
Inchi Key CAXRNOBCSUDRMU-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436451

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4193_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart