KEY428075881 Key Organics / BIONET

5-bromo-N-tert-butylpyridine-3-sulfonamide

  • MDL number MFCD16103444
  • Cas Number : 911111-80-3
  • Molecular Weight : 293.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436452

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4195_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart