KEY428075885 Key Organics / BIONET

7-bromo-2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

  • MDL number MFCD02168781
  • Cas Number : 3951-89-1
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436456

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4201_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart