KEY428075893 Key Organics / BIONET

(3-chloropyridin-2-yl)methanamine dihydrochloride

  • MDL number MFCD07371473
  • Cas Number : 342816-31-3
  • Molecular Weight : 215.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436464

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4218_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart