KEY428075897 Key Organics / BIONET

6-benzyl-1,2,3,4,5,6,7,8-octahydro-1,6-naphthyridin-2-one

  • MDL number MFCD09403131
  • Cas Number : 210539-03-0
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436468

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4360_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart