KEY428075899 Key Organics / BIONET

tert-butyl N-cyclopropylcarbamate

  • MDL number MFCD09701238
  • Cas Number : 132844-48-5
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl cyclopropylcarbamate
Inchi Code 1S/C8H15NO2/c1-8(2,3)11-7(10)9-6-4-5-6/h6H,4-5H2,1-3H3,(H,9,10)
Inchi Key WFFGQLYKMWGXMU-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436470

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4362_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart