KEY428075914 Key Organics / BIONET

(3Z)-3-[(2-iodophenyl)methylidene]-1-methyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD04535810
  • Cas Number : 1616502-83-0
  • Molecular Weight : 361.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436485

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4387_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart