KEY428075923 Key Organics / BIONET

{5-[(acetyloxy)methyl]-1,4-dioxan-2-yl}methyl acetate

  • MDL number MFCD28098226
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4405_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart