KEY428075924 Key Organics / BIONET

methyl 4-cyano-3-fluorobenzoate

  • MDL number MFCD09264509
  • Cas Number : 268734-34-5
  • Molecular Weight : 179.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-cyano-3-fluorobenzoate
Inchi Code 1S/C9H6FNO2/c1-13-9(12)6-2-3-7(5-11)8(10)4-6/h2-4H,1H3
Inchi Key AHJLHWRWDRCIKO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436495

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4406_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart