KEY428075931 Key Organics / BIONET

5-iodo-1,3-benzoxazole-2-thiol

  • MDL number MFCD09261223
  • Cas Number : 93614-44-9
  • Molecular Weight : 277.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436502

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4417_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart