KEY428075941 Key Organics / BIONET

methyl (2Z)-2-acetamido-3-(4-hydroxy-2,6-dimethylphenyl)prop-2-enoate

  • MDL number MFCD28098230
  • Cas Number : 1616502-84-1
  • Molecular Weight : 263.293

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222 - 224
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DK-0730_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart