KEY428075957 Key Organics / BIONET

ethyl 2-oxo-1,2-dihydroquinoline-3-carboxylate

  • MDL number MFCD00572653
  • Cas Number : 85870-47-9
  • Molecular Weight : 217.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 162 (Lit)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436528

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EK-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart