KEY428075958 Key Organics / BIONET

methyl (2Z)-2-acetamido-3-(2,3,6-trifluorophenyl)prop-2-enoate

  • MDL number MFCD28098238
  • Cas Number : 1616502-86-3
  • Molecular Weight : 273.211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129 - 131
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EK-0723_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart