KEY428075969 Key Organics / BIONET

methyl (2Z)-3-[4-(dimethylamino)phenyl]-2-acetamidoprop-2-enoate

  • MDL number MFCD28098241
  • Cas Number : 944261-89-6
  • Molecular Weight : 262.309

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 165
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436540

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FK-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart