KEY428075972 Key Organics / BIONET

1-(3-ethoxy-4-methoxyphenyl)-2-methanesulfonylethan-1-amine

  • MDL number MFCD27665207
  • Cas Number : 253168-94-4
  • Molecular Weight : 273.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436543

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FK-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart