KEY428075974 Key Organics / BIONET

N-{2-[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-methanesulfonylethyl]-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl}acetamide

  • MDL number MFCD18782607
  • Cas Number : 608141-41-9
  • Molecular Weight : 460.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436545

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FK-0727_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart