KEY428075980 Key Organics / BIONET

3-(bromomethyl)-8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD28098243
  • Cas Number : 1616500-70-9
  • Molecular Weight : 314.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436551

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GF-0014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart