KEY428075984 Key Organics / BIONET

(cyclopentylideneamino)thiourea

  • MDL number MFCD00458163
  • Cas Number : 7283-39-8
  • Molecular Weight : 157.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-155
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436583

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MS-11550_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart