KEY428075987 Key Organics / BIONET

4H-imidazo[4,5,-i,j]-quinolin-2,6-(1H,5H)dione

  • MDL number MFCD24566098
  • Cas Number : 75655-10-6
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436586

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart