KEY428075989 Key Organics / BIONET

4-(2-((3-fluoro-5-(trifluoromethyl)phenyl)sulfonyl)propan-2-yl)piperidine HCL

  • MDL number MFCD20482272
  • Cas Number : 1245784-26-2
  • Molecular Weight : 389.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436588

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4806_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart