KEY428075991 Key Organics / BIONET

benzyl 2-{[(tert-butoxy)carbonyl]amino}acetate

  • MDL number MFCD09032588
  • Cas Number : 54244-69-8
  • Molecular Weight : 265.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436590

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart