KEY428075992 Key Organics / BIONET

(R)-2-amino-2-(2-chlorophenyl)acetamide hydrochloride

  • MDL number MFCD28098245
  • Cas Number : 374629-84-2
  • Molecular Weight : 221.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436591

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4809_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart