KEY428075995 Key Organics / BIONET

(3-fluoro-5-methoxypyridin-2-yl)methanol

  • MDL number MFCD16611254
  • Cas Number : 1227581-08-9
  • Molecular Weight : 157.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436594

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4812_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart