KEY428075997 Key Organics / BIONET

S-methyl-N-(tert-butyldimethylsilyl)-S-phenylsulfoximine

  • MDL number MFCD28098246
  • Cas Number : 117893-43-3
  • Molecular Weight : 269.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436596

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4814_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart