KEY428075998 Key Organics / BIONET

(5-{2,4-dichloropyrido[2,3-d]pyrimidin-7-yl}-2-methoxyphenyl)methyl 2,2-dimethylpropanoate

  • MDL number MFCD18378252
  • Cas Number : 1201798-97-1
  • Molecular Weight : 420.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436597

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart