KEY428076001 Key Organics / BIONET

methyl 5-cyclopropyl-3-(2,6-dichlorophenyl)-1,2-oxazole-4-carboxylate

  • MDL number MFCD12964218
  • Cas Number : 946426-88-6
  • Molecular Weight : 312.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436600

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4818_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart