KEY428412235 Key Organics / BIONET

2-methoxyethyl N-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamoyl}carbamate

  • MDL number MFCD28101486
  • Molecular Weight : 364.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436918

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2842_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart