KEY428412239 Key Organics / BIONET

potassium (4-butanamidophenyl)trifluoroboranuide

  • MDL number MFCD28101490
  • Molecular Weight : 269.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name potassium (4-butyramidophenyl)trifluoroborate
Inchi Code 1S/C10H12BF3NO.K/c1-2-3-10(16)15-9-6-4-8(5-7-9)11(12,13)14;/h4-7H,2-3H2,1H3,(H,15,16);/q-1;+1
Inchi Key QVSNOOWDNZKDSE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436926

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2987_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart