KEY428412241 Key Organics / BIONET

1-{4-[(1S,2S,6R,8S)-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0^{2,6}]decan-4-yl]phenyl}ethan-1-one

  • MDL number MFCD28101492
  • Molecular Weight : 298.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2990_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart