KEY428412242 Key Organics / BIONET

(1S,2S,6R,8S)-2,9,9-trimethyl-4-(2-nitrophenyl)-3,5-dioxa-4-boratricyclo[6.1.1.0^{2,6}]decane

  • MDL number MFCD28101493
  • Molecular Weight : 301.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436932

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2993_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart