KEY428412252 Key Organics / BIONET

4,4,6-trimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane

  • MDL number MFCD28101503
  • Cas Number : 1092060-77-9
  • Molecular Weight : 218.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436958

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart