KEY428412254 Key Organics / BIONET

2-(4-iodophenyl)-4,4,6-trimethyl-1,3,2-dioxaborinane

  • MDL number MFCD28101505
  • Cas Number : 1279115-53-5
  • Molecular Weight : 329.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436960

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart