KEY428412258 Key Organics / BIONET

4-(4,4,6-trimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile

  • MDL number MFCD28101509
  • Cas Number : 1092060-81-5
  • Molecular Weight : 229.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436968

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart