KEY428412263 Key Organics / BIONET

potassium [4-(acetyloxy)-3,5-bis(propan-2-yl)phenyl]trifluoroboranuide

  • MDL number MFCD28101513
  • Molecular Weight : 326.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436975

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3036_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart