KEY428412266 Key Organics / BIONET

tetrabutylazanium; trifluoro(4-fluorophenyl)boranuide

  • MDL number MFCD28101518
  • Cas Number : 1291068-40-0
  • Molecular Weight : 405.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436987

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart