KEY428412275 Key Organics / BIONET

2-(4-fluorophenyl)-4,4,6-trimethyl-1,3,2-dioxaborinane

  • MDL number MFCD08457859
  • Cas Number : 1029653-69-7
  • Molecular Weight : 222.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-fluorophenyl)-4,4,6-trimethyl-1,3,2-dioxaborinane
Inchi Code 1S/C12H16BFO2/c1-9-8-12(2,3)16-13(15-9)10-4-6-11(14)7-5-10/h4-7,9H,8H2,1-3H3
Inchi Key ARFOLXGRXVAHCE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436955

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3016_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart