KEY428412286 Key Organics / BIONET

diethyl({[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine

  • MDL number MFCD09746207
  • Cas Number : 1012785-46-4
  • Molecular Weight : 289.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]ethanamine
Inchi Code 1S/C17H28BNO2/c1-7-19(8-2)13-14-11-9-10-12-15(14)18-20-16(3,4)17(5,6)21-18/h9-12H,7-8,13H2,1-6H3
Inchi Key ZSIJHVXJNJQXDF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436966

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3027_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart