KEY428412303 Key Organics / BIONET

4-bromo-2-(morpholin-4-yl)benzonitrile

  • MDL number MFCD11037539
  • Cas Number : 1260762-06-8
  • Molecular Weight : 267.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436944

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart