KEY428412317 Key Organics / BIONET

5-bromopyrimidine-2,4-diamine

  • MDL number MFCD11848482
  • Cas Number : 1004-01-9
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,4-pyrimidinediamine
Inchi Code 1S/C4H5BrN4/c5-2-1-8-4(7)9-3(2)6/h1H,(H4,6,7,8,9)
Inchi Key SUHXPSAYNUCJSC-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436996

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4147_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart