KEY428412321 Key Organics / BIONET

2-fluoro-4-hydroxy-5-nitrobenzonitrile

  • MDL number MFCD16658599
  • Cas Number : 1134198-38-1
  • Molecular Weight : 182.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437000

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4151_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart