KEY428412336 Key Organics / BIONET

5-(benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid

  • MDL number MFCD08531038
  • Cas Number : 1219-33-6
  • Molecular Weight : 246.218

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437015

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4175_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart