KEY428412354 Key Organics / BIONET

2-chloro-4-(piperidin-4-yloxy)benzonitrile hydrochloride

  • MDL number MFCD28101528
  • Cas Number : 1620482-39-4
  • Molecular Weight : 273.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437033

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4253_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart